Anti-CD1A Rabbit Monoclonal Antibody [clone: EP3622]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM1013-C1 ILM1013-C01 ILM1013-C05
ILM1013-C1EA 1770 EUR
ILM1013-C1 IMMLILM1013-C01 IMMLILM1013-C05
Anti-CD1A Rabbit Monoclonal Antibody [clone: EP3622]
Antilichamen
Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: EP3622
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR