Anti-CR2 Rabbit Monoclonal Antibody [clone: EP3093]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM4004-C01 ILM4004-C05 ILM4004-C1
ILM4004-C01EA 189 EUR
ILM4004-C01 ILM4004-C05 ILM4004-C1
Anti-CR2 Rabbit Monoclonal Antibody [clone: EP3093]
Antilichamen
Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: EP3093
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR