Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: BER H2]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM1113-C01 ILM1113-C05 ILM1113-C1
ILM1113-C01EA 69.3 EUR
ILM1113-C01 ILM1113-C05 ILM1113-C1
Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: BER H2]
Antilichamen
Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: BER H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human