Anti-SIGLEC6 Mouse Monoclonal Antibody [clone: PWS44]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM1303-C01 ILM1303-C05 ILM1303-C1
ILM1303-C01EA 296 EUR
ILM1303-C01 ILM1303-C05 ILM1303-C1
Anti-SIGLEC6 Mouse Monoclonal Antibody [clone: PWS44]
Antilichamen
Type: Primary
Antigen: SIGLEC6
Clonality: Monoclonal
Clone: PWS44
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgGb2
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR