Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND/10]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM1343-C01 ILM1343-C05 ILM1343-C1
ILM1343-C01EA 78.8 EUR
ILM1343-C01 ILM1343-C05 ILM1343-C1
Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND/10]
Antilichamen
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: QBEND/10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Cynomolgus Monkey, Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR