Anti-SPN Mouse Monoclonal Antibody [clone: DF-T1]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM6693-C01 ILM6693-C05 ILM6693-C1
ILM6693-C01EA 78.8 EUR
ILM6693-C01 ILM6693-C05 ILM6693-C1
Anti-SPN Mouse Monoclonal Antibody [clone: DF-T1]
Antilichamen
Type: Primary
Antigen: SPN
Clonality: Monoclonal
Clone: DF-T1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR