Anti-NCAM1 Mouse Monoclonal Antibody [clone: 123C3]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM15611-C01 ILM15611-C05 ILM15611-C1
ILM15611-C01EA 78.8 EUR
ILM15611-C01 ILM15611-C05 ILM15611-C1
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 123C3]
Antilichamen
Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 123C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human