Anti-ITGB3 Mouse Monoclonal Antibody [clone: 2F2]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM1603-C01 ILM1603-C05 ILM1603-C1
ILM1603-C01EA 146 EUR
ILM1603-C01 ILM1603-C05 ILM1603-C1
Anti-ITGB3 Mouse Monoclonal Antibody [clone: 2F2]
Antilichamen
Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: 2F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR