Anti-CD68 Mouse Monoclonal Antibody [clone: KP1]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM9689-C01 ILM9689-C05 ILM9689-C1
ILM9689-C01EA 101 EUR
ILM9689-C01 ILM9689-C05 ILM9689-C1
Anti-CD68 Mouse Monoclonal Antibody [clone: KP1]
Antilichamen
Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: KP1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Rabbit, Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR