Anti-CD7 Rabbit Monoclonal Antibody [clone: EP132]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM3007-C01 ILM3007-C05 ILM3007-C1
ILM3007-C01EA 238 EUR
ILM3007-C01 ILM3007-C05 ILM3007-C1
Anti-CD7 Rabbit Monoclonal Antibody [clone: EP132]
Antilichamen
Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: EP132
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human