Anti-CD8A Mouse Monoclonal Antibody [clone: C8/144B]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM1083-C01 ILM1083-C05 ILM1083-C1
ILM1083-C01EA 99.6 EUR
ILM1083-C01 ILM1083-C05 ILM1083-C1
Anti-CD8A Mouse Monoclonal Antibody [clone: C8/144B]
Antilichamen
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: C8/144B
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR