Anti-CD99 Mouse Monoclonal Antibody [clone: 12E7]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM4267-C01 ILM4267-C05 ILM4267-C1
ILM4267-C01EA 96.6 EUR
ILM4267-C01 ILM4267-C05 ILM4267-C1
Anti-CD99 Mouse Monoclonal Antibody [clone: 12E7]
Antilichamen
Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: 12E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR