Anti-KIT Rabbit Monoclonal Antibody [clone: YR145]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM7683-C01 ILM7683-C05 ILM7683-C1
ILM7683-C01EA 159 EUR
ILM7683-C01 ILM7683-C05 ILM7683-C1
Anti-KIT Rabbit Monoclonal Antibody [clone: YR145]
Antilichamen
Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: YR145
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR