Anti-CDX2 Rabbit Monoclonal Antibody [clone: EPR2764Y]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM2353-R10 ILM2353-R25
VWRKILM2353-R10EA 0 EUR
VWRKILM2353-R10 VWRKILM2353-R25
Anti-CDX2 Rabbit Monoclonal Antibody [clone: EPR2764Y]
Antilichamen
Type: Primary
Antigen: CDX2
Clonality: Monoclonal
Clone: EPR2764Y
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human