Anti-KRT8/KRT18 Mouse Monoclonal Antibody [clone: K8.8+DC10]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM6910-C1 ILM6910-C01 ILM6910-C05
ILM6910-C1EA 495 EUR
ILM6910-C1 ILM6910-C01 ILM6910-C05
Anti-KRT8/KRT18 Mouse Monoclonal Antibody [clone: K8.8+DC10]
Antilichamen
Type: Primary
Antigen: KRT8/KRT18
Clonality: Monoclonal
Clone: K8.8+DC10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR