Anti-KRT19 Mouse Monoclonal Antibody [clone: A53-B/A2.26]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM3880-C01 ILM3880-C05
ILM3880-C01EA 78.8 EUR
ILM3880-C01 ILM3880-C05
Anti-KRT19 Mouse Monoclonal Antibody [clone: A53-B/A2.26]
Antilichamen
Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: A53-B/A2.26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR