Anti-Cytokeratin Mouse Monoclonal Antibody [clone: 34BETAE12]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM3343-C01 ILM3343-C05 ILM3343-C1
ILM3343-C01EA 112 EUR
ILM3343-C01 ILM3343-C05 ILM3343-C1
Anti-Cytokeratin Mouse Monoclonal Antibody [clone: 34BETAE12]
Antilichamen
Type: Primary
Antigen: Cytokeratin
Clonality: Monoclonal
Clone: 34BETAE12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR