Anti-Cytokeratin Mouse Monoclonal Antibody [clone: AE1]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM0253-C01 ILM0253-C05 ILM0253-C1
ILM0253-C01EA 63 EUR
ILM0253-C01 ILM0253-C05 ILM0253-C1
Anti-Cytokeratin Mouse Monoclonal Antibody [clone: AE1]
Antilichamen
Type: Primary
Antigen: Cytokeratin
Clonality: Monoclonal
Clone: AE1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Cow, Dog, Rabbit, Mouse, Rat, Chicken, Turtle
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR