Anti-CDH1 Mouse Monoclonal Antibody [clone: MX020]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM7002-C01 ILM7002-C05 ILM7002-C1
ILM7002-C01EA 211 EUR
ILM7002-C01 ILM7002-C05 ILM7002-C1
Anti-CDH1 Mouse Monoclonal Antibody [clone: MX020]
Antilichamen
Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone: MX020
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human