Anti-MUC1 Mouse Monoclonal Antibody [clone: GP1.4]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM4529-C01 ILM4529-C05 ILM4529-C1
ILM4529-C01EA 78.8 EUR
ILM4529-C01 ILM4529-C05 ILM4529-C1
Anti-MUC1 Mouse Monoclonal Antibody [clone: GP1.4]
Antilichamen
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: GP1.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human