Anti-FGFR1 Rat Monoclonal Antibody [clone: ER-TR7]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM76619-C01 ILM76619-C05 ILM76619-C1
ILM76619-C01EA 194 EUR
ILM76619-C01 ILM76619-C05 ILM76619-C1
Anti-FGFR1 Rat Monoclonal Antibody [clone: ER-TR7]
Antilichamen
Type: Primary
Antigen: FGFR1
Clonality: Monoclonal
Clone: ER-TR7
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR