Anti-GFAP Rabbit Monoclonal Antibody [clone: EP672Y]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM3353-C01 ILM3353-C05 ILM3353-C1
ILM3353-C01EA 161 EUR
ILM3353-C01 ILM3353-C05 ILM3353-C1
Anti-GFAP Rabbit Monoclonal Antibody [clone: EP672Y]
Antilichamen
Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: EP672Y
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR