Anti-G. lamblia Goat Polyclonal Antibody

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILP62210-C01 ILP62210-C05 ILP62210-C1
ILP62210-C01EA 121 EUR
ILP62210-C01 ILP62210-C05 ILP62210-C1
Anti-G. lamblia Goat Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: G. lamblia
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Whole Organism
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR