Anti-GPC3 Mouse Monoclonal Antibody [clone: 1G12]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM2613-C01 ILM2613-C05 ILM2613-C1
ILM2613-C01EA 186 EUR
ILM2613-C01 ILM2613-C05 ILM2613-C1
Anti-GPC3 Mouse Monoclonal Antibody [clone: 1G12]
Antilichamen
Type: Primary
Antigen: GPC3
Clonality: Monoclonal
Clone: 1G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR