Anti-LM02 Mouse Monoclonal Antibody [clone: 1A9-1]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM2400-C01 ILM2400-C05 ILM2400-C1
ILM2400-C01EA 184 EUR
ILM2400-C01 ILM2400-C05 ILM2400-C1
Anti-LM02 Mouse Monoclonal Antibody [clone: 1A9-1]
Antilichamen
Type: Primary
Antigen: LM02
Clonality: Monoclonal
Clone: 1A9-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat