Anti-MITF Mouse Monoclonal Antibody [clone: 34CA5]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM1056-C01 ILM1056-C05 ILM1056-C1
ILM1056-C01EA 318 EUR
ILM1056-C01 ILM1056-C05 ILM1056-C1
Anti-MITF Mouse Monoclonal Antibody [clone: 34CA5]
Antilichamen
Type: Primary
Antigen: MITF
Clonality: Monoclonal
Clone: 34CA5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR