Anti-MITF Mouse Monoclonal Antibody [clone: C5/D5]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM2843-C01 ILM2843-C05 ILM2843-C1
ILM2843-C01EA 142 EUR
ILM2843-C01 ILM2843-C05 ILM2843-C1
Anti-MITF Mouse Monoclonal Antibody [clone: C5/D5]
Antilichamen
Type: Primary
Antigen: MITF
Clonality: Monoclonal
Clone: Clone : C5/D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 & IgG1
Reactivity: Human