Anti-MYOD1 Mouse Monoclonal Antibody [clone: 5.8A]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM4456-C01 ILM4456-C05 ILM4456-C1
ILM4456-C01EA 78.8 EUR
ILM4456-C01 ILM4456-C05 ILM4456-C1
Anti-MYOD1 Mouse Monoclonal Antibody [clone: 5.8A]
Antilichamen
Type: Primary
Antigen: MYOD1
Clonality: Monoclonal
Clone: Clone : 5.8A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR