Anti-NAPSA Mouse Monoclonal Antibody [clone: NAPSA/1239]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM9479-C1 ILM9479-C01 ILM9479-C05
ILM9479-C1EA 795 EUR
ILM9479-C1 ILM9479-C01 ILM9479-C05
Anti-NAPSA Mouse Monoclonal Antibody [clone: NAPSA/1239]
Antilichamen
Type: Primary
Antigen: NAPSA
Clonality: Monoclonal
Clone: NAPSA/1239
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human