Anti-CDKN2A Mouse Monoclonal Antibody [clone: G175-405]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM5620-C01 ILM5620-C05 ILM5620-C1
ILM5620-C01EA 91.2 EUR
ILM5620-C01 ILM5620-C05 ILM5620-C1
Anti-CDKN2A Mouse Monoclonal Antibody [clone: G175-405]
Antilichamen
Type: Primary
Antigen: CDKN2A
Clonality: Monoclonal
Clone: G175-405
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human