Anti-CDKN1C Mouse Monoclonal Antibody [clone: KP10]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM4573-C01 ILM4573-C05 ILM4573-C1
ILM4573-C01EA 134 EUR
ILM4573-C01 ILM4573-C05 ILM4573-C1
Anti-CDKN1C Mouse Monoclonal Antibody [clone: KP10]
Antilichamen
Type: Primary
Antigen: CDKN1C
Clonality: Monoclonal
Clone: KP10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR