Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 15D2]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM89619-C01 ILM89619-C05 ILM89619-C1
ILM89619-C01EA 195 EUR
ILM89619-C01 ILM89619-C05 ILM89619-C1
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 15D2]
Antilichamen
Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 15D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Canine