Anti-PAX5 Mouse Monoclonal Antibody [clone: MX017]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM0706-C01 ILM0706-C05 ILM0706-C1
ILM0706-C01EA 233 EUR
ILM0706-C01 ILM0706-C05 ILM0706-C1
Anti-PAX5 Mouse Monoclonal Antibody [clone: MX017]
Antilichamen
Type: Primary
Antigen: PAX5
Clonality: Monoclonal
Clone: MX017
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR