Anti-PDPN Mouse Monoclonal Antibody [clone: D2-40]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM4174-C01 ILM4174-C05 ILM4174-C1
ILM4174-C01EA 244 EUR
ILM4174-C01 ILM4174-C05 ILM4174-C1
Anti-PDPN Mouse Monoclonal Antibody [clone: D2-40]
Antilichamen
Type: Primary
Antigen: PDPN
Clonality: Monoclonal
Clone: D2-40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR