Anti-PGR Rabbit Monoclonal Antibody [clone: ZR4]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM0222-C05 ILM0222-C1 ILM0222-C01
ILM0222-C05EA 945 EUR
ILM0222-C05 ILM0222-C1 IMMLILM0222-C01
Anti-PGR Rabbit Monoclonal Antibody [clone: ZR4]
Antilichamen
Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: ZR4
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human