Anti-CA9 Mouse Monoclonal Antibody [clone: PN-15]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM3293-C01 ILM3293-C05 ILM3293-C1
ILM3293-C01EA 88.9 EUR
ILM3293-C01 ILM3293-C05 ILM3293-C1
Anti-CA9 Mouse Monoclonal Antibody [clone: PN-15]
Antilichamen
Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: PN-15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Horse