Anti-STAT6 Mouse Monoclonal Antibody [clone: D1]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM3719-C01 ILM3719-C05 ILM3719-C1
ILM3719-C01EA 162 EUR
ILM3719-C01 ILM3719-C05 ILM3719-C1
Anti-STAT6 Mouse Monoclonal Antibody [clone: D1]
Antilichamen
Type: Primary
Antigen: STAT6
Clonality: Monoclonal
Clone: D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR