Anti-TG Mouse Monoclonal Antibody [clone: 2H11 + 6E1]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM1133-C01 ILM1133-C05 ILM1133-C1
ILM1133-C01EA 98.9 EUR
ILM1133-C01 ILM1133-C05 ILM1133-C1
Anti-TG Mouse Monoclonal Antibody [clone: 2H11 + 6E1]
Antilichamen
Type: Primary
Antigen: TG
Clonality: Monoclonal
Clone: 2H11 + 6E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR