Anti-TPSAB1 Mouse Monoclonal Antibody [clone: G3]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM3423-C01 ILM3423-C05 ILM3423-C1
ILM3423-C01EA 99.6 EUR
ILM3423-C01 ILM3423-C05 ILM3423-C1
Anti-TPSAB1 Mouse Monoclonal Antibody [clone: G3]
Antilichamen
Type: Primary
Antigen: TPSAB1
Clonality: Monoclonal
Clone: G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR