Anti-NKX2-1 Mouse Monoclonal Antibody [clone: SPT24]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM105846-C01 ILM105846-C05 ILM105846-C1
ILM105846-C01EA 159 EUR
ILM105846-C01 ILM105846-C05 ILM105846-C1
Anti-NKX2-1 Mouse Monoclonal Antibody [clone: SPT24]
Antilichamen
Type: Primary
Antigen: NKX2-1
Clonality: Monoclonal
Clone: SPT24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR