Anti-TYR Mouse Monoclonal Antibody [clone: T311]

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ILM7299-C01 ILM7299-C05 ILM7299-C1
ILM7299-C01EA 63 EUR
ILM7299-C01 ILM7299-C05 ILM7299-C1
Anti-TYR Mouse Monoclonal Antibody [clone: T311]
Antilichamen
Type: Primary
Antigen: TYR
Clonality: Monoclonal
Clone: T311
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR