Anti-VIM Mouse Monoclonal Antibody [clone: V9]
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILM52311-C01 ILM52311-C05 ILM52311-C1
ILM52311-C01EA 125 EUR
ILM52311-C01 ILM52311-C05 ILM52311-C1
Anti-VIM Mouse Monoclonal Antibody [clone: V9]
Antilichamen
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: V9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human