Precision cut thermometers
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4736854 4736855 4736856
SCNL4736854EA 89.2 EUR
SCNL4736854 SCNL4736855 SCNL4736856
Precision cut thermometers
Thermometers Analoge thermometers
These standard grinding size precision cut thermometers are designed with handle.
  • Colourless, round capillary form
Upper part: 275×11 mm
Lower part: Ø 7,25 mm