Pasteur pipettes, disposable, Jumbo Pastettes®
Leverancier: Alpha Laboratories
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LW4263 LW4264 LW4266
612-1706EA 88.7 EUR
612-1706 ALPCLW4264 ALPCLW4266
Pasteur pipettes, disposable, Jumbo Pastettes®
Pipetten Pasteur-/transferpipetten
Manufactured from low density polyethylene (LDPE). With a soft and responsive bulb, Jumbo Pastettes provide a high level of dexterity and control for quick and easy use.
  • Perfect for accessing deep vessels such as Winchesters and tissue culture flasks
  • Can be recycled if used with non hazardous samples
Jumbo Pastettes are versatile and safe to use with a wide variety of liquids including oils, making them suitable for a whole host of applications from tissue culture to engineering.