pH buffer solutions, certified (DKD)
Leverancier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
51350046 51350044 51350042 51350038 51350036 51350034 51350032 51350048
METT51350046EA 94.25 EUR
METT51350046 METT51350044 METT51350042 METT51350038 METT51350036 METT51350034 METT51350032 METT51350048
pH buffer solutions, certified (DKD)
Buffers pH-buffers
Certificaten: The DKD (Deutscher Kalibrierdienst, German Calibration Service) certified buffers are the ideally suitable products for regulated industries. DKD is accepted by other accreditation bodies within the scope of the European Cooperation for Accreditation (EA) and the International Laboratory Accreditation Cooperation (ILAC).