Acid special cleaner, TICKOPUR R 27
Leverancier: BANDELIN-ELECTRONIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
815 816 874
142-1940EA 47.5 EUR
142-1940 142-1941 142-1943
Acid special cleaner, TICKOPUR R 27
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Special cleaner based on phosphoric acid, for decalcification and rust removal with corrosion protection.