Conductivity standards, potassium chloride solutions
Leverancier: CONSORT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B560 B561 B562
CONSB560EA 26.9 EUR
CONSB560 CONSB561 CONSB562
Conductivity standards, potassium chloride solutions
Standaarden Geleidbaarheidsstandaarden
Potassium chloride solutions referenced at 25 °C.
  • Certified to 0,5%