Capsule counting trays, SteriWare®
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8444A-1 8444A-1S
SAMP8444A-1EA 107 EUR
SAMP8444A-1 SAMP8444A-1S
Capsule counting trays, SteriWare®
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
The disposable counting trays are made from robust, shatterproof PS. They are suitable for use with all capsules from size 2 to 00.
  • BSE/TSE free
  • Suitable for capsules size 2 to size 00
  • Manufactured and packed in a cleanroom
  • Can be used straight from the bag
Certificaten: Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.
Verpakking: Individually bagged in a heat-sealed laminate bag.