Alkaline cleaner, TICKOPUR R 60
Leverancier: BANDELIN-ELECTRONIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
818 896
142-1961EA 50.5 EUR
142-1961 142-1986
Alkaline cleaner, TICKOPUR R 60
Reinigingsmiddelen Detergenten en oppervlaktereinigers
TICKOPUR R 60 is an intensive, phosphate-free, strongly alkaline cleaner. These can be used to clean steel, stainless steel, glass, ceramics, plastics and rubber.