Antibody diluent (HAMA blocker) for ELISA
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB193969-50 AB193969-12
ABCAAB193969-50EA 280 EUR
ABCAAB193969-50 ABCAAB193969-12
Antibody diluent (HAMA blocker) for ELISA
Buffers Buffers voor zuivering van antilichamen
Antibody diluent (HAMA blocker) for ELISA is a proprietary reagent for immunoassays involving a sandwich antibody complex that are susceptible to heterophilic antibody interference.
  • Store at +4 °C
These diluents are specially formulated to quantitate your proteins of interest in complex sample types, such as human serum or human plasma. Formulation is shown to alleviate antibody interference due to heterophilic antibodies such as human anti-mouse antibody (HAMA). These antibody diluents (HAMA blocker) for ELISA is shown to eliminate heterophilic/HAMA interferences without compromising true signal quantitation.
Opgelet:
All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.